siRNA Details
| Gene Symbol: Nxnl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001271341.1 | NP_001258270.1 | 1 | 266 | ||
| XM_006252893.2 | XP_006252955.1 | 1 | 751 | |||
| XM_006252894.2 | XP_006252956.1 | 1 | 751 | |||
| XM_008771104.1 | XP_008769326.1 | 1 | 751 | |||
| GenBank mRNA | EU861029.1 | - | 1 | 278 | ||

Nxnl1
Genomic Map