siRNA Details
| Gene Symbol: Armc6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107306.1 | NP_001100776.1 | 2 | 425 | ||
| GenBank mRNA | BC166821.1 | - | 81 | 8368184 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Armc6| Gene Symbol: Armc6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107306.1 | NP_001100776.1 | 2 | 425 | ||
| GenBank mRNA | BC166821.1 | - | 81 | 8368184 | ||