siRNA Details
| Gene Symbol: Pragmin | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107315.1 | NP_001100785.1 | 3, 4 | 2334 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pragmin| Gene Symbol: Pragmin | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107315.1 | NP_001100785.1 | 3, 4 | 2334 | ||