siRNA Details
| Gene Symbol: Proz | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006253473.1 | XP_006253535.1 | 7 | 792 | ||
| XM_006253474.1 | XP_006253536.1 | 6 | 678 | |||
| GenBank mRNA | BC160868.1 | - | 7 | 704 | ||
| FQ209855.1 | - | 7 | 706 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Proz| Gene Symbol: Proz | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006253473.1 | XP_006253535.1 | 7 | 792 | ||
| XM_006253474.1 | XP_006253536.1 | 6 | 678 | |||
| GenBank mRNA | BC160868.1 | - | 7 | 704 | ||
| FQ209855.1 | - | 7 | 706 | |||