siRNA Details
| Gene Symbol: Ing1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001038591.1 | NP_001033680.1 | 1 | 358 | ||
| GenBank mRNA | AY624101.1 | - | 1 | 358 | ||
| AY624102.1 | - | 1 | 358 | |||
| AY624103.1 | - | 278 | ||||
| AY624104.1 | - | 278 | ||||
| AY624105.1 | - | 2 | 285 | |||
| BC166594.1 | - | 351 | 7937717 | |||

Ing1
Genomic Map