siRNA Details
Gene Symbol: Spata31d3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008771468.1 | XP_008769690.1 | 4 | 1014 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Spata31d3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008771468.1 | XP_008769690.1 | 4 | 1014 |