siRNA Details
Gene Symbol: Cxcl14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013137.2 | NP_001013155.1 | 3, 4 | 476 | ||
GenBank mRNA | BC101896.1 | - | 3, 4 | 476 | 7932648 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Cxcl14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013137.2 | NP_001013155.1 | 3, 4 | 476 | ||
GenBank mRNA | BC101896.1 | - | 3, 4 | 476 | 7932648 |