siRNA Details
| Gene Symbol: Tspan17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013138.2 | NP_001013156.2 | 8 | 1046 | ||
| GenBank mRNA | AY485933.1 | - | 8 | 1018 | ||
| BC097431.1 | - | 8 | 1026 | 7455325 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tspan17| Gene Symbol: Tspan17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013138.2 | NP_001013156.2 | 8 | 1046 | ||
| GenBank mRNA | AY485933.1 | - | 8 | 1018 | ||
| BC097431.1 | - | 8 | 1026 | 7455325 | ||