siRNA Details
| Gene Symbol: Sema4d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001170563.1 | NP_001164034.1 | 5 | 911 | ||
| GenBank mRNA | DN935876.1 | - | 2 | 208 | ||
| FQ211736.1 | - | 1128 | ||||
| FQ212166.1 | - | 1249 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Sema4d| Gene Symbol: Sema4d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001170563.1 | NP_001164034.1 | 5 | 911 | ||
| GenBank mRNA | DN935876.1 | - | 2 | 208 | ||
| FQ211736.1 | - | 1128 | ||||
| FQ212166.1 | - | 1249 | ||||