siRNA Details
| Gene Symbol: Ippk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008556.1 | NP_001008556.1 | 13 | 1355 | ||
| XM_008771534.1 | XP_008769756.1 | 12 | 2070 | |||
| XM_008771535.1 | XP_008769757.1 | 13 | 1454 | |||
| GenBank mRNA | AY823319.1 | - | 13 | 1355 | ||

Ippk
Genomic Map