siRNA Details
| Gene Symbol: Zfp169 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006253745.2 | XP_006253807.1 | 5 | 1514 | ||
| XM_225207.8 | XP_225207.5 | 5 | 1511 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zfp169| Gene Symbol: Zfp169 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006253745.2 | XP_006253807.1 | 5 | 1514 | ||
| XM_225207.8 | XP_225207.5 | 5 | 1511 | |||