siRNA Details
| Gene Symbol: LOC103689985 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006253865.2 | XP_006253927.1 | 6 | 1347 | ||
| XM_008771601.1 | XP_008769823.1 | 6 | 1347 | |||
| Gene Symbol: Mylk4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006253874.2 | XP_006253936.1 | 7 | 1610 | ||

LOC103689985
Genomic Map