siRNA Details
| Gene Symbol: Fam65b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014009.2 | NP_001014031.2 | 18 | 2286 | ||
| XM_006253929.2 | XP_006253991.1 | 15 | 2715 | |||
| XM_006253930.1 | XP_006253992.1 | 15 | 2071 | |||
| XM_008771636.1 | XP_008769858.1 | 14 | 2429 | |||
| XM_008771637.1 | XP_008769859.1 | 14 | 1991 | |||
| GenBank mRNA | AY325194.1 | - | 18 | 2286 | ||
| FQ226921.1 | - | 2 | 117 | |||

Fam65b
Genomic Map