siRNA Details
| Gene Symbol: Slc17a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107353.1 | NP_001100823.1 | 11 | 1683 | ||
| XM_006253969.2 | XP_006254031.1 | 11 | 1760 | |||
| XM_008771656.1 | XP_008769878.1 | 11 | 2944 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Slc17a2| Gene Symbol: Slc17a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107353.1 | NP_001100823.1 | 11 | 1683 | ||
| XM_006253969.2 | XP_006254031.1 | 11 | 1760 | |||
| XM_008771656.1 | XP_008769878.1 | 11 | 2944 | |||