siRNA Details
| Gene Symbol: Zfp192 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001100574.1 | NP_001094044.1 | 3 | 743 | ||
| XM_008771718.1 | XP_008769940.1 | 3 | 1054 | |||
| XM_008771719.1 | XP_008769941.1 | 3 | 750 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zfp192| Gene Symbol: Zfp192 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001100574.1 | NP_001094044.1 | 3 | 743 | ||
| XM_008771718.1 | XP_008769940.1 | 3 | 1054 | |||
| XM_008771719.1 | XP_008769941.1 | 3 | 750 | |||