siRNA Details
Gene Symbol: Adnp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127373.1 | NP_001120845.1 | 4 | 631 | ||
GenBank mRNA | BC166606.1 | - | 4 | 631 | 7934474 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Adnp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127373.1 | NP_001120845.1 | 4 | 631 | ||
GenBank mRNA | BC166606.1 | - | 4 | 631 | 7934474 |