siRNA Details
| Gene Symbol: Prdm6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008772189.1 | XP_008770411.1 | 6 | 1472 | ||
| XM_008772190.1 | XP_008770412.1 | 5 | 1746 | |||
| XM_008772191.1 | XP_008770413.1 | 6 | 1472 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Prdm6| Gene Symbol: Prdm6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008772189.1 | XP_008770411.1 | 6 | 1472 | ||
| XM_008772190.1 | XP_008770412.1 | 5 | 1746 | |||
| XM_008772191.1 | XP_008770413.1 | 6 | 1472 | |||