siRNA Details
| Gene Symbol: Zfp532 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107382.1 | NP_001100852.1 | 7 | 3148 | ||
| XM_006254865.2 | XP_006254927.1 | 7 | 5438 | |||
| XM_006254866.2 | XP_006254928.1 | 7 | 4877 | |||
| XM_006254868.2 | XP_006254930.1 | 7 | 3465 | |||
| XM_006254869.2 | XP_006254931.1 | 7 | 3310 | |||
| XM_006254870.2 | XP_006254932.1 | 7 | 4842 | |||
| XM_006254871.2 | XP_006254933.1 | 6 | 3023 | |||
| XM_006254872.2 | XP_006254934.1 | 7 | 4222 | |||
| XM_006254873.2 | XP_006254935.1 | 6 | 5328 | |||
| XM_006254875.2 | XP_006254937.1 | 3 | 3542 | |||
| XM_008772142.1 | XP_008770364.1 | 6 | 4064 | |||

Zfp532
Genomic Map