siRNA Details
| Gene Symbol: Napg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107384.1 | NP_001100854.1 | 5, 6 | 481 | ||
| XM_006254829.2 | XP_006254891.1 | 6, 7 | 473 | |||
| GenBank mRNA | AY724541.1 | - | 6, 7 | 378 | ||
| FQ211390.1 | - | 3, 4 | 355 | |||

Napg
Genomic Map