siRNA Details
| Gene Symbol: Napg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107384.1 | NP_001100854.1 | 7 | 577 | ||
| XM_006254829.2 | XP_006254891.1 | 8 | 569 | |||
| GenBank mRNA | AY724541.1 | - | 8 | 474 | ||
| FQ211390.1 | - | 5 | 451 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Napg| Gene Symbol: Napg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107384.1 | NP_001100854.1 | 7 | 577 | ||
| XM_006254829.2 | XP_006254891.1 | 8 | 569 | |||
| GenBank mRNA | AY724541.1 | - | 8 | 474 | ||
| FQ211390.1 | - | 5 | 451 | |||