siRNA Details
| Gene Symbol: Tnfaip8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107387.1 | NP_001100857.1 | 4 | 917 | ||
| XM_006254708.2 | XP_006254770.1 | 2 | 6829 | |||
| XM_006254709.2 | XP_006254771.1 | 2 | 569 | |||
| XM_008772149.1 | XP_008770371.1 | 2 | 2907 | |||
| GenBank mRNA | FQ230976.1 | - | 2 | 587 | ||
| FQ235239.1 | - | 2 | 587 | |||

Tnfaip8
Genomic Map