siRNA Details
| Gene Symbol: Tcerg1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107390.2 | NP_001100860.2 | 19 | 2783 | ||
| XM_006254673.2 | XP_006254735.1 | 20 | 2837 | |||
| XM_006254674.2 | XP_006254736.1 | 19 | 2774 | |||
| XM_006254675.2 | XP_006254737.1 | 18 | 2723 | |||
| GenBank mRNA | BC168862.1 | - | 19 | 2743 | 7444403 | |

Tcerg1
Genomic Map