siRNA Details
| Gene Symbol: Gpr151 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_181633.2 | NP_853664.1 | 1 | 1097 | ||
| GenBank mRNA | AJ564168.1 | - | 1 | 1097 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gpr151| Gene Symbol: Gpr151 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_181633.2 | NP_853664.1 | 1 | 1097 | ||
| GenBank mRNA | AJ564168.1 | - | 1 | 1097 | ||
Set Membership: |
|