siRNA Details
| Gene Symbol: Hars2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014012.2 | NP_001014034.2 | 10 | 1127 | ||
| XM_006254575.2 | XP_006254637.1 | 9 | 1037 | |||
| GenBank mRNA | BC089973.1 | - | 3 | 291 | 7316402 | |
| CV075096.1 | - | 7 | 730 | |||

Hars2
Genomic Map