siRNA Details
| Gene Symbol: Papd5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107416.1 | NP_001100886.1 | 10 | 1410 | ||
| XM_006255261.1 | XP_006255323.1 | 12 | 2020 | |||
| XM_008772403.1 | XP_008770625.1 | 13 | 2031 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Papd5| Gene Symbol: Papd5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107416.1 | NP_001100886.1 | 10 | 1410 | ||
| XM_006255261.1 | XP_006255323.1 | 12 | 2020 | |||
| XM_008772403.1 | XP_008770625.1 | 13 | 2031 | |||