siRNA Details
| Gene Symbol: Jph3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107437.1 | NP_001100907.1 | 2 | 817 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Jph3| Gene Symbol: Jph3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107437.1 | NP_001100907.1 | 2 | 817 | ||