siRNA Details
| Gene Symbol: Ablim1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008760582.1 | XP_008758804.1 | 12, 13 | 1718 | ||
| GenBank mRNA | BC105867.1 | - | 5, 6 | 568 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ablim1| Gene Symbol: Ablim1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008760582.1 | XP_008758804.1 | 12, 13 | 1718 | ||
| GenBank mRNA | BC105867.1 | - | 5, 6 | 568 | ||