siRNA Details
| Gene Symbol: Mcoln3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012059.1 | NP_001012059.1 | 13 | 1710 | ||
| GenBank mRNA | BC079200.1 | - | 13 | 1710 | 7131704 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mcoln3| Gene Symbol: Mcoln3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012059.1 | NP_001012059.1 | 13 | 1710 | ||
| GenBank mRNA | BC079200.1 | - | 13 | 1710 | 7131704 | |