siRNA Details
| Gene Symbol: Slc2a12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107451.1 | NP_001100921.1 | 2 | 1585 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Slc2a12| Gene Symbol: Slc2a12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107451.1 | NP_001100921.1 | 2 | 1585 | ||