siRNA Details
| Gene Symbol: Nkd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107454.1 | NP_001100924.1 | 10 | 1108 | ||
| XM_008758691.1 | XP_008756913.1 | 6 | 1806 | |||
| XM_008758692.1 | XP_008756914.1 | 13 | 2842 | |||
| XM_008758693.1 | XP_008756915.1 | 10 | 1623 | |||

Nkd2
Genomic Map