siRNA Details
| Gene Symbol: Fndc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001038615.1 | NP_001033704.1 | 8 | 1252 | ||
| XM_006227871.2 | XP_006227933.1 | 11 | 1757 | |||
| XM_008758732.1 | XP_008756954.1 | 10 | 1568 | |||
| XM_008758733.1 | XP_008756955.1 | 8 | 1406 | |||
| GenBank mRNA | DQ256268.1 | - | 8 | 1252 | ||

Fndc1
Genomic Map