siRNA Details
| Gene Symbol: Acat2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001006995.1 | NP_001006996.1 | 7 | 836 | ||
| GenBank mRNA | AY325187.1 | - | 810 | |||
| BC083872.1 | - | 7 | 836 | 7128564 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Acat2| Gene Symbol: Acat2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001006995.1 | NP_001006996.1 | 7 | 836 | ||
| GenBank mRNA | AY325187.1 | - | 810 | |||
| BC083872.1 | - | 7 | 836 | 7128564 | ||