siRNA Details
| Gene Symbol: Kptn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107457.1 | NP_001100927.1 | 1 | 149 | ||
| GenBank mRNA | BC166768.1 | - | 121 | 7933544 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Kptn| Gene Symbol: Kptn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107457.1 | NP_001100927.1 | 1 | 149 | ||
| GenBank mRNA | BC166768.1 | - | 121 | 7933544 | ||