siRNA Details
| Gene Symbol: Zfp541 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003748775.3 | XP_003748823.1 | 7 | 3112 | ||
| XM_006228357.2 | XP_006228419.1 | 8 | 3177 | |||
| XM_006228358.2 | XP_006228420.1 | 7 | 2849 | |||
| XM_008759027.1 | XP_008757249.1 | 9 | 3287 | |||
| XM_008759028.1 | XP_008757250.1 | 7 | 2896 | |||
| XM_008759029.1 | XP_008757251.1 | 8 | 3222 | |||

Zfp541
Genomic Map