siRNA Details
| Gene Symbol: Cnksr3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012061.1 | NP_001012061.1 | 3 | 798 | ||
| XM_006227875.2 | XP_006227937.1 | 3 | 1955 | |||
| GenBank mRNA | AY328890.1 | - | 3 | 798 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cnksr3| Gene Symbol: Cnksr3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012061.1 | NP_001012061.1 | 3 | 798 | ||
| XM_006227875.2 | XP_006227937.1 | 3 | 1955 | |||
| GenBank mRNA | AY328890.1 | - | 3 | 798 | ||