siRNA Details
| Gene Symbol: LOC102547059 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006228753.2 | XP_006228815.1 | 10 | 3053 | ||
| XM_006228754.2 | XP_006228816.1 | 10 | 3053 | |||
| XM_008759228.1 | XP_008757450.1 | 9 | 2259 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC102547059| Gene Symbol: LOC102547059 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006228753.2 | XP_006228815.1 | 10 | 3053 | ||
| XM_006228754.2 | XP_006228816.1 | 10 | 3053 | |||
| XM_008759228.1 | XP_008757450.1 | 9 | 2259 | |||