siRNA Details
Gene Symbol: Uba2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100579.1 | NP_001094049.1 | 11 | 1250 | ||
GenBank mRNA | BC161985.1 | - | 11 | 1250 | 9030809 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Uba2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100579.1 | NP_001094049.1 | 11 | 1250 | ||
GenBank mRNA | BC161985.1 | - | 11 | 1250 | 9030809 |