siRNA Details
| Gene Symbol: Cpt1c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001034925.2 | NP_001030097.2 | 11, 12 | 1544 | ||
| XM_006229066.2 | XP_006229128.1 | 11, 12 | 2566 | |||
| XM_008759376.1 | XP_008757598.1 | 10, 11 | 2710 | |||
| XM_008759377.1 | XP_008757599.1 | 9, 10 | 1547 | |||
| GenBank mRNA | BC105882.1 | - | 2, 3 | 307 | ||

Cpt1c
Genomic Map