siRNA Details
| Gene Symbol: Sec1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001135584.1 | NP_001129056.1 | 1 | 571 | ||
| XM_006229074.2 | XP_006229136.1 | 6 | 1195 | |||
| XM_008759379.1 | XP_008757601.1 | 2 | 760 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Sec1| Gene Symbol: Sec1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001135584.1 | NP_001129056.1 | 1 | 571 | ||
| XM_006229074.2 | XP_006229136.1 | 6 | 1195 | |||
| XM_008759379.1 | XP_008757601.1 | 2 | 760 | |||