siRNA Details
| Gene Symbol: Ndn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008558.1 | NP_001008558.1 | 1 | 992 | ||
| GenBank mRNA | BC087630.1 | - | 1 | 992 | 7319134 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ndn| Gene Symbol: Ndn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008558.1 | NP_001008558.1 | 1 | 992 | ||
| GenBank mRNA | BC087630.1 | - | 1 | 992 | 7319134 | |