siRNA Details
| Gene Symbol: Prc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107529.1 | NP_001100999.1 | 12 | 1621 | ||
| XM_006229433.2 | XP_006229495.1 | 12 | 1633 | |||
| XM_006229437.2 | XP_006229499.1 | 12 | 1633 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Prc1| Gene Symbol: Prc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107529.1 | NP_001100999.1 | 12 | 1621 | ||
| XM_006229433.2 | XP_006229495.1 | 12 | 1633 | |||
| XM_006229437.2 | XP_006229499.1 | 12 | 1633 | |||