siRNA Details
| Gene Symbol: Det1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001037194.1 | NP_001032271.1 | 4 | 1589 | ||
| GenBank mRNA | BC100253.1 | - | 4 | 1589 | 7456473 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Det1| Gene Symbol: Det1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001037194.1 | NP_001032271.1 | 4 | 1589 | ||
| GenBank mRNA | BC100253.1 | - | 4 | 1589 | 7456473 | |