siRNA Details
| Gene Symbol: Adamtsl3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107533.2 | NP_001101003.2 | 7 | 961 | ||
| XM_006229477.2 | XP_006229539.1 | 7 | 956 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Adamtsl3| Gene Symbol: Adamtsl3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107533.2 | NP_001101003.2 | 7 | 961 | ||
| XM_006229477.2 | XP_006229539.1 | 7 | 956 | |||