siRNA Details
Gene Symbol: Adamtsl3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107533.2 | NP_001101003.2 | 19 | 2641 | ||
XM_006229477.2 | XP_006229539.1 | 19 | 2636 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Adamtsl3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107533.2 | NP_001101003.2 | 19 | 2641 | ||
XM_006229477.2 | XP_006229539.1 | 19 | 2636 |