siRNA Details
| Gene Symbol: Prss23 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001007691.1 | NP_001007692.1 | 2 | 808 | ||
| GenBank mRNA | BC079179.1 | - | 2 | 808 | 7130532 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Prss23| Gene Symbol: Prss23 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001007691.1 | NP_001007692.1 | 2 | 808 | ||
| GenBank mRNA | BC079179.1 | - | 2 | 808 | 7130532 | |