siRNA Details
| Gene Symbol: Tsku | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009965.2 | NP_001009965.1 | 2 | 982 | ||
| XM_006229759.2 | XP_006229821.1 | 2 | 994 | |||
| XM_006229760.2 | XP_006229822.1 | 2 | 1875 | |||
| XM_006229761.2 | XP_006229823.1 | 2 | 1177 | |||
| GenBank mRNA | AY533242.1 | - | 2 | 903 | ||
| BC100644.1 | - | 2 | 982 | 7930712 | ||

Tsku
Genomic Map