siRNA Details
| Gene Symbol: Usp47 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107542.2 | NP_001101012.2 | 10 | 1505 | ||
| XM_006230044.2 | XP_006230106.1 | 11 | 1389 | |||
| XM_006230045.2 | XP_006230107.1 | 10 | 1569 | |||
| XM_006230046.2 | XP_006230108.1 | 11 | 1389 | |||
| GenBank mRNA | BC166609.1 | - | 10 | 1505 | ||

Usp47
Genomic Map