siRNA Details
Gene Symbol: Dchs1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107544.2 | NP_001101014.2 | 4 | 2463 | ||
XM_006229942.2 | XP_006230004.1 | 5 | 2652 | |||
XM_008759732.1 | XP_008757954.1 | 5 | 5374 |
