siRNA Details
| Gene Symbol: Tmem219 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003748988.3 | XP_003749036.1 | 7 | 1372 | ||
| XM_008759928.1 | XP_008758150.1 | 6 | 1058 | |||
| XM_008759929.1 | XP_008758151.1 | 5 | 863 | |||
| XM_008759930.1 | XP_008758152.1 | 5 | 860 | |||
| GenBank mRNA | BC166576.1 | - | 6 | 1954 | 7321704 | |
| FQ216353.1 | - | 3 | 770 | |||

Tmem219
Genomic Map