siRNA Details
| Gene Symbol: Mcmbp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001039608.2 | NP_001034697.2 | 14 | 1956 | ||
| GenBank mRNA | BC111072.1 | - | 1 | 200 | 7445757 | |
| BC160884.1 | - | 14 | 1956 | 7383399 | ||
| FQ225204.1 | - | 5 | 662 | |||

Mcmbp
Genomic Map